3-Amino-2-methylpyridine

Synonyms: 2-Methylpyridin-3-amine, 2-Methyl-3-pyridylamine, 3-Amino-2-picoline

662690-250MG 662690-1G
220085-801EA 79.37 USD
220085-801 220085-800
3-Amino-2-methylpyridine
3-Amino-2-methylpyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD03788195
CAS Number: 3430-10-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR