3,4-Dimethoxythiophenol ≥97%

Warning

Synonyms: 3,4-Dimethoxybenzene-1-thiol, 3,4-Dimethoxybenzenethiol, 4-Mercaptoveratrole, 3,4-Dimethoxyphenyl mercaptan, 1,2-Dimethoxy-4-mercaptobenzene

662518-10G 662518-1G
220086-718EA 56.18 USD
220086-718 220086-719
3,4-Dimethoxythiophenol ≥97%
3,4-Dimethoxythiophenol
Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
Boiling Pt: 86 - 88 °C (0.4 -rr)
Density: 1.19 g/cm³ (20 °C)
MDL Number: MFCD00052823
CAS Number: 700-96-9
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR