Propyltrimethoxysilane

Synonyms: Trimethoxy(propyl)silane

662275-1L 662275-100ML
220086-694EA 98.45 USD
220086-694 220086-693
Propyltrimethoxysilane
Propyltrimethoxysilane
Formula: C₆H₁₆O₃Si
MW: 164.28 g/mol
Boiling Pt: 142 - 143 °C (1013 hPa)
Density: 0.935 g/cm³ (20 °C)
MDL Number: MFCD00043026
CAS Number: 1067-25-0
EINECS: 213-926-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR