3-Cyano-5-methylpyridine

Synonyms: 3-Cyano-5-picoline, 5-Methylpyridine-3-carbonitrile, 5-Methylnicotinonitrile

661929-1G 661929-250MG
220084-330EA 110.92 USD
220084-330 220086-662
3-Cyano-5-methylpyridine
3-Cyano-5-methylpyridine
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 220 °C (1013 hPa)
Melting Pt: 86 °C
MDL Number: MFCD00673151
CAS Number: 42885-14-3
EINECS: 255-988-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR