4-Bromo-o-anisaldehyde ≥97%

Synonyms: 4-Bromo-2-methoxybenzaldehyde

661880-5G 661880-1G
220084-323EA 253.85 USD
220084-323 220086-661
4-Bromo-o-anisaldehyde ≥97%
4-Bromo-o-anisaldehyde
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD08276823
CAS Number: 43192-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR