2-Bromo-5-(trifluoromethyl)pyridine

661120-5G 661120-1G
220086-614EA 239.08 USD
220086-614 220086-613
2-Bromo-5-(trifluoromethyl)pyridine
2-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
MW: 226 g/mol
MDL Number: MFCD00153086
CAS Number: 50488-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR