4-Amino-3-(trifluoromethyl)pyridine

661090-500MG 661090-100MG
220086-608EA 303.69 USD
220086-608 220086-607
4-Amino-3-(trifluoromethyl)pyridine
4-Amino-3-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD02183554
CAS Number: 387824-61-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR