2-Amino-6-(trifluoromethyl)pyridine

660574-500MG 660574-100MG
220086-581EA 228.46 USD
220086-581 220086-580
2-Amino-6-(trifluoromethyl)pyridine
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 78…80 °C
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR