2-Amino-4-(trifluoromethyl)pyridine

660345-100MG 660345-500MG
220086-558EA 147.69 USD
220086-558 220086-559
2-Amino-4-(trifluoromethyl)pyridine
2-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD00077455
CAS Number: 106447-97-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR