2-Fluoro-5-iodopyridine

660043-10G 660043-1G
220086-530EA 212.15 USD
220086-530 220086-531
2-Fluoro-5-iodopyridine
2-Fluoro-5-iodopyridine
Formula: C₅H₃FIN
MW: 222.99 g/mol
MDL Number: MFCD03095301
CAS Number: 171197-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR