Potassium(3-fluorophenyl)trifluoroborate ≥96%

Synonyms: Potassium trifluoro(3-fluorophenyl)borate, (3-Fluorophenyl)trifluoroboric acid potassium salt

659770-5G 659770-1G
220086-062EA 353.08 USD
220086-062 220086-061
Potassium(3-fluorophenyl)trifluoroborate ≥96%
Potassium(3-fluorophenyl)trifluoroborate
Formula: C₆H₄BF₄K
MW: 202 g/mol
MDL Number: MFCD06659938
CAS Number: 267006-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR