3-Iodo-4-nitrotoluene ≥97%

659320-5G 659320-25G
220084-315EA 108.77 USD
220084-315 220084-314
3-Iodo-4-nitrotoluene ≥97%
3-Iodo-4-nitrotoluene
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 95 - 97 °C
CAS Number: 52488-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR