4-Bromo-3-iodotoluene ≥97%

659312-5G 659312-1G
220084-313EA 126 USD
220084-313 220086-029
4-Bromo-3-iodotoluene ≥97%
4-Bromo-3-iodotoluene
Formula: C₇H₆BrI
MW: 296.93 g/mol
MDL Number: MFCD07784420
CAS Number: 858841-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR