3-Bromo-4-nitrobenzoic acid ≥97%

Warning

659304-1G 659304-5G
220086-027EA 70 USD
220086-027 220086-028
3-Bromo-4-nitrobenzoic acid ≥97%
3-Bromo-4-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
MDL Number: MFCD04117949
CAS Number: 101420-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR