4-Hydroxy-3-methylbenzoic acid ≥97%

Synonyms: 4-Hydroxy-m-toluic acid

659282-10G 659282-1G
220086-462EA 127.08 USD
220086-462 220086-463
4-Hydroxy-3-methylbenzoic acid ≥97%
4-Hydroxy-3-methylbenzoic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00270105
CAS Number: 499-76-3
EINECS: 207-890-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR