Os EnCat® 40

Danger

658685-1G 658685-500MG
220086-432EA 94.15 USD
220086-432 220086-195
Os EnCat® 40
Osmium tetraoxide

Os EnCat® 40 is osmium tetroxide encapsulated within a porous cross-linked polyurea matrix.


  • extent of labeling: 0.3 mmol/g Os loading


Encapsulated OsO4 catalyst offers safety and handling advantages over conventional OsO4

Formula: O₄Os
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: 40 °C
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011150
CAS Number: 20816-12-0
EINECS: 244-058-7
UN: 2471
ADR: 6.1,I
Merck Index: 13,06962

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR