2,4-Dichloro-5-nitropyrimidine

Warning

658340-50G 658340-1G 658340-10G
220085-436EA 756 USD
220085-436 220085-435 220085-434
2,4-Dichloro-5-nitropyrimidine
2,4-Dichloro-5-nitropyrimidine
Formula: C₄HCl₂N₃O₂
MW: 193.98 g/mol
Boiling Pt: 326 °C (1013 hPa)
Melting Pt: 29.3 °C
Density: 1.74 g/cm³ (20 °C)
MDL Number: MFCD00127867
CAS Number: 49845-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR