2,6-Dichloronicotinic acid

Synonyms: 2,6-Dichloro-3-pyridinecarboxylic acid, 2,6-Dichloropyridine-3-carboxylic acid

658219-25G 658219-5G
220086-319EA 122.77 USD
220086-319 220086-320
2,6-Dichloronicotinic acid
2,6-Dichloronicotinic acid
Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
MDL Number: MFCD00075583
CAS Number: 38496-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR