2-Methyl-9,10-anthraquinone, Technical Grade

Synonyms: 2-Methylanthraquinone, 2-Methylanthracene-9,10-Dione

65800-500G 65800-100G
220086-291EA 269.23 USD
220086-291 220086-290
2-Methyl-9,10-anthraquinone, Technical Grade
2-Methyl-9,10-anthraquinone
Formula: C₁₅H₁₀O₂
MW: 222.24 g/mol
Boiling Pt: 236 - 238 °C (10 -rr)
Melting Pt: 171 - 173 °C
Density: 1.365 g/cm³ (20 °C)
MDL Number: MFCD00001235
CAS Number: 84-54-8
EINECS: 201-539-6
UN: 3077
ADR: 9,III
Merck Index: 12,06100

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR