2-Chloro-3-methoxybenzoic acid ≥97%

Synonyms: 2-Chloro-m-anisic acid

657905-10G 657905-1G
220086-280EA 103.83 USD
220086-280 220086-281
2-Chloro-3-methoxybenzoic acid ≥97%
2-Chloro-3-methoxybenzoic acid
Formula: C₈H₇ClO₃
MW: 186.59 g/mol
MDL Number: MFCD07784399
CAS Number: 33234-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR