4-Amino-3-iodobenzonitrile ≥98%

Synonyms: 4-Cyano-2-iodoaniline

657123-5G 657123-1G
220086-002EA 116.31 USD
220086-002 220086-001
4-Amino-3-iodobenzonitrile ≥98%
4-Amino-3-iodobenzonitrile
Formula: C₇H₅IN₂
MW: 244.03 g/mol
MDL Number: MFCD04039964
CAS Number: 33348-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR