1,2,3-Trifluoro-5-nitrobenzene ≥97%

Danger

Synonyms: 1,2,3-Trifluoro-5-nitrobenzene, 3,4,5-Trifluoronitrobenzene

656917-5G 656917-25G
220085-771EA 159.38 USD
220085-771 220085-770
1,2,3-Trifluoro-5-nitrobenzene ≥97%
1,2,3-Trifluoro-5-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
MDL Number: MFCD00456803
CAS Number: 66684-58-0
EINECS: 266-447-0
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR