3-(2,4-Dichlorophenyl)propionic acid

Synonyms: 3-(2,4-Dichlorophenyl)propanoic acid

656186-5G 656186-1G
220085-884EA 66.46 USD
220085-884 220085-883
3-(2,4-Dichlorophenyl)propionic acid
3-(2,4-Dichlorophenyl)propionic acid
Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD00275605
CAS Number: 55144-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR