3-(m-Tolyl)propionic acid

Synonyms: 3-(3-Methylphenyl)propionic acid, 3-(3-Methylphenyl)propanoic acid

656178-5G 656178-1G
220085-882EA 200.31 USD
220085-882 220085-881
3-(m-Tolyl)propionic acid
3-(m-Tolyl)propionic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00060326
CAS Number: 3751-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR