3-(4-Chlorophenyl)propionic acid

656151-1G 656151-5G
220085-879EA 42.32 USD
220085-879 220085-880
3-(4-Chlorophenyl)propionic acid
3-(4-Chlorophenyl)propionic acid
Formula: C₉H₉ClO₂
MDL Number: MFCD00016555
CAS Number: 2019-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR