2,4-Dichloro-6-(trifluoromethyl)aniline ≥97%

Warning

Synonyms: 2-Amino-3,5-dichloro-α,α,α-trifluorotoluene, 4,6-Dichloro-α,α,α-trifluoro-o-toluidine

656011-1G 656011-5G
220085-857EA 82.92 USD
220085-857 220085-858
2,4-Dichloro-6-(trifluoromethyl)aniline ≥97%
2,4-Dichloro-6-(trifluoromethyl)aniline
Formula: C₇H₄Cl₂F₃N
MW: 230.02 g/mol
MDL Number: MFCD00174313
CAS Number: 62593-17-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR