3-Bromo-4,5-diaminobenzotrifluoride ≥97%

Synonyms: 3-Bromo-4,5-diamino-α,α,α-trifluorotoluene, 3-Bromo-5-(trifluoromethyl)-1,2-phenylenediamine, 1-Bromo-2,3-diamino-5-(trifluoromethyl)benzene, 3-Bromo-5-(trifluoromethyl)-o-phenylenediamine, 3-Bromo-5-(trifluoromethyl)benzene-1,2-diamine

656003-1G 656003-5G
220085-855EA 39.52 USD
220085-855 220085-856
3-Bromo-4,5-diaminobenzotrifluoride ≥97%
3-Bromo-4,5-diaminobenzotrifluoride
Formula: C₇H₆BrF₃N₂
MW: 255.04 g/mol
MDL Number: MFCD00042484
CAS Number: 113170-72-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR