4-Bromo-2-methyl-6-nitroaniline ≥97%

Synonyms: 4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene

655988-1G 655988-5G
220085-852EA 570 USD
220085-852 220085-853
4-Bromo-2-methyl-6-nitroaniline ≥97%
4-Bromo-2-methyl-6-nitroaniline
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00052919
CAS Number: 77811-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR