2-Bromo-6-methyl-4-nitroaniline ≥97%

Synonyms: 2-Amino-3-bromo-5-nitrotoluene, 6-Bromo-4-nitro-o-toluidine

655937-5G 655937-1G
220085-851EA 570 USD
220085-851 220085-850
2-Bromo-6-methyl-4-nitroaniline ≥97%
2-Bromo-6-methyl-4-nitroaniline
Formula: C₇H₇BrN₂O₂
MW: 231.05 g/mol
MDL Number: MFCD00053089
CAS Number: 102170-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR