3-(3-Fluorophenyl)propionic acid ≥97%

Synonyms: 3-(3-Fluorophenyl)propanoic acid

655252-5G 655252-1G
220085-721EA 77.54 USD
220085-721 220085-720
3-(3-Fluorophenyl)propionic acid ≥97%
3-(3-Fluorophenyl)propionic acid
Formula: C₉H₉FO₂
MW: 168.17 g/mol
MDL Number: MFCD01310821
CAS Number: 458-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR