1-(2,3-Difluorophenyl)-2-thiourea ≥97%

Synonyms: N-(2,3-Difluorophenyl)thiourea, 1-(2,3-Difluorophenyl)thiourea, 2,3-Difluorophenylthiourea

654833-1G 654833-5G
220085-662EA 83.89 USD
220085-662 220085-663
1-(2,3-Difluorophenyl)-2-thiourea ≥97%
1-(2,3-Difluorophenyl)-2-thiourea
Formula: (C₆H₃F₂)NC(S)NH₂
MW: 188.2 g/mol
MDL Number: MFCD07368705
CAS Number: 572889-25-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR