1-(3-Fluorophenyl)-2-thiourea ≥97%

Danger

Synonyms: N-(3-Fluorophenyl)thiourea, 3-Fluorophenylthiourea

654825-1G 654825-5G
220085-660EA 45.98 USD
220085-660 220085-661
1-(3-Fluorophenyl)-2-thiourea ≥97%
1-(3-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
Boiling Pt: 276 °C (1013 hPa)
Melting Pt: 116 °C
MDL Number: MFCD00041167
CAS Number: 458-05-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR