Nickel(II) chloride hexahydrate

Danger

Synonyms: Nickel dichloride hexahydrate

654507-25G 654507-5G 654507-100G
220085-623EA 141.38 USD
220085-623 220085-624 220085-622
Nickel(II) chloride hexahydrate
Nickel(II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR