4-Iodo-2-nitroaniline ≥97%

654108-10G 654108-1G
220085-585EA 285 USD
220085-585 220085-586
4-Iodo-2-nitroaniline ≥97%
4-Iodo-2-nitroaniline
Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD06808526
CAS Number: 20691-72-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR