3-Amino-4-chlorobenzonitrile ≥96%

653837-10G 653837-1G
220084-310EA 171.92 USD
220084-310 220084-311
3-Amino-4-chlorobenzonitrile ≥96%
3-Amino-4-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
MDL Number: MFCD00017089
CAS Number: 53312-79-1
EINECS: 258-471-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR