4-Bromo-2-methoxybenzyl alcohol ≥97%

653187-1G 653187-10G
220085-464EA 32.2 USD
220085-464 220085-463
4-Bromo-2-methoxybenzyl alcohol ≥97%
4-Bromo-2-methoxybenzyl alcohol
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
MDL Number: MFCD07369762
CAS Number: 17102-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR