4-Hydroxy-o-toluic acid ≥98%

Synonyms: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid

653160-1G 653160-10G
220085-461EA 67.85 USD
220085-461 220085-460
4-Hydroxy-o-toluic acid ≥98%
4-Hydroxy-o-toluic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD02182261
CAS Number: 578-39-2
EINECS: 209-422-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR