3-Methoxypropiononitrile ≥98.0% (by GC)

Warning

Synonyms: 3-Methoxypropanenitrile, 3-Methoxypropionitrile

65290-1L-F 65290-250ML-F
220085-447EA 249.85 USD
220085-447 220085-448
3-Methoxypropiononitrile ≥98.0% (by GC)
3-Methoxypropiononitrile
Formula: C₄H₇NO
MW: 85.11 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.94 g/cm³ (20 °C)
MDL Number: MFCD00001958
CAS Number: 110-67-8
EINECS: 203-790-7

Order Now

Specification Test Results

Assay 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR