2-Chloro-4-iodoaniline ≥97%

652679-5G 652679-25G
220085-381EA 116.31 USD
220085-381 220085-380
2-Chloro-4-iodoaniline ≥97%
2-Chloro-4-iodoaniline
Formula: C₆H₅ClIN
MW: 253.47 g/mol
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 73 °C
MDL Number: MFCD00025298
CAS Number: 42016-93-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR