2-Bromo-5-iodopyridine

Warning

652598-5G 652598-25G
220085-370EA 103.17 USD
220085-370 220085-369
2-Bromo-5-iodopyridine
2-Bromo-5-iodopyridine
Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD03095201
CAS Number: 73290-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR