3-Bromo-4-hydroxybenzonitrile ≥97%

652563-5G 652563-25G
220085-366EA 66.92 USD
220085-366 220085-365
3-Bromo-4-hydroxybenzonitrile ≥97%
3-Bromo-4-hydroxybenzonitrile
Formula: C₇H₄BrNO
MW: 198.02 g/mol
Boiling Pt: 406 °C (1013 hPa)
Melting Pt: 155 °C
MDL Number: MFCD00143096
CAS Number: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR