Barium fluoride ≥99.999%

Warning

652458-25G 652458-100G
220085-354EA 131.38 USD
220085-354 220085-353
Barium fluoride ≥99.999%
Barium fluoride
Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1270 °C
Density: 4.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 12,01003

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR