3,4,5-Trimethoxyphenylboronic acid

Warning

Synonyms: 3,4,5-Trimethoxybenzeneboronic acid, Dihydroxy(3,4,5-trimethoxyphenyl)borane, (3,4,5-Trimethoxyphenyl)metaboric acid

651648-5G 651648-25G
220082-431EA 489.08 USD
220082-431 220085-259
3,4,5-Trimethoxyphenylboronic acid
3,4,5-Trimethoxyphenylboronic acid
Formula: C₉H₁₃BO₅
MW: 212.01 g/mol
MDL Number: MFCD01075676
CAS Number: 182163-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR