4-Amino-3-nitrophenylboronic acid pinacol ester ≥97%

Synonyms: 4-Amino-3-nitrophenylboronic Acid Pinacol Ester, 4-Amino-3-nitrophenylboronic acid, pinacol ester

651613-5G 651613-1G
220085-257EA 200.77 USD
220085-257 220085-256
4-Amino-3-nitrophenylboronic acid pinacol ester ≥97%
4-Amino-3-nitrophenylboronic acid pinacol ester
Formula: C₁₂H₁₇BN₂O₄
MW: 264.09 g/mol
MDL Number: MFCD06795680
CAS Number: 833486-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR