2-Amino-5-chloropyrimidine

Synonyms: 5-Chloropyrimidin-2-ylamine, 5-Chloro-2-pyrimidinamine, 5-Chlor-2-pyrimidinamin, 5-chlorpyrimidin-2-amin

651567-1G 651567-5G
220085-250EA 91 USD
220085-250 220085-251
2-Amino-5-chloropyrimidine
2-Amino-5-chloropyrimidine
Formula: C₄H₄ClN₃
MW: 129.55 g/mol
MDL Number: MFCD00014604
CAS Number: 5428-89-7
EINECS: 226-581-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR