1-Boc-3-piperidone

Synonyms: N-BOC-3-piperidone, 1-Boc-3-Piperidinone

650811-25G 650811-5G
220085-170EA 83.57 USD
220085-170 220085-171
1-Boc-3-piperidone
1-Boc-3-piperidone
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
MDL Number: MFCD01631193
CAS Number: 98977-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR