4-Amino-2-fluorobenzoic acid ≥97%

650625-250MG 650625-1G
220084-261EA 79.37 USD
220084-261 220084-260
4-Amino-2-fluorobenzoic acid ≥97%
4-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD01569397
CAS Number: 446-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR