Hexane (mixture of isomers) ≥98.5%, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®

Danger

Synonyms: Hexanes, Isohexane

650544-1L 650544-4X4L 650544-6X1L 650544-4L
220084-580EA 37.23 USD
220084-580 220084-583 220084-584 220084-582
Hexane (mixture of isomers) ≥98.5%, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68 - 70 °C (1013 hPa)
Melting Pt: -95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: -22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR