2,3-Dichlorothiophene

650293-1G 650293-10G
220085-134EA 29.62 USD
220085-134 220085-133
2,3-Dichlorothiophene
2,3-Dichlorothiophene
Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 169…170 °C (1013 hPa)
Melting Pt: –26.2 °C
Density: 1.4605 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00043890
CAS Number: 17249-79-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR