4,5-Dichloro-2-thiophenesulfonyl chloride

Synonyms: 4,5-Dichlorothiophene-2-sulphonyl chloride, 2,3-Dichloro-5-thiophenesulphonyl chloride, 2,3-Dichlorothiophene-5-sulphonyl chloride

650285-1G 650285-10G
220085-132EA 299.08 USD
220085-132 220085-131
4,5-Dichloro-2-thiophenesulfonyl chloride
4,5-Dichloro-2-thiophenesulphonyl chloride
Formula: C₄HCl₃O₂S₂
MW: 251.54 g/mol
Melting Pt: 56 °C
MDL Number: MFCD00052092
CAS Number: 126714-85-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR